Isopropylamine, Sigma-Aldrich®
numer katalogowy SIAL59330-1L-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:59.11 g/mol
Merck Index:13,05229
Boiling Pt:32.4 °C (1013 hPa)
Melting Pt:–101 °C
Density:0.69 g/cm³ (20 °C)
Flash Pt:–27 °C
Storage Temperature:Ambient
EINECS:200-860-9
UN:1221
ADR:3,I
:GHS02
:GHS07
CAS Number:75-31-0
Formula:C₃H₉N
MDL Number:MFCD00008082