


1,2-Dichloroethane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-06-2
:
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963
:98.96 g/mol
Merck Index:13,03831
Boiling Pt:83.5 °C (1013 hPa)
Melting Pt:–35.5 °C
Density:1.254 g/cm³ (20 °C)
EINECS:203-458-1
UN:1184
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:13 °C
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together