Order Entry
Poland
ContactUsLinkComponent
 
 
 
Acetic acid 99-100% for synthesis, Sigma-Aldrich®
 :  Merck
 
undefined
 
 
Acetic acid 99-100% for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    17 °C
  • Molar Mass:
    60.05 g/mol
  • Boiling Point:
    116 - 118 °C (1013 hPa)
  • Vapor Pressure:
    15.4 hPa (20 °C)
  • Flash Point:
    44 °C
  • Refractive Index:
    1.37 (20 °C)
  • Explosion Limit:
    4 - 19.9 %(V)
  • Density:
    1.05 g/cm3 (20 °C)
  • pH:
    2.5 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    485 °C
  • Saturation concentration:
    38 g/m3 (20 °C) Air
  • Viscosity kinematic:
    1.17 mm2/s (20 °C)
  • Evaporation number:
    24
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

Frequently Bought Together