Order Entry
Poland
ContactUsLinkComponent
Capric acid for synthesis, Sigma-Aldrich®
Irritant
Capric acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Capric acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.15 g/l (20 °C)
  • Melting Point:
    29 - 32 °C
  • Molar Mass:
    172.26 g/mol
  • Bulk Density:
    690 kg/m3
  • Boiling Point:
    268 - 270 °C
  • Vapor Pressure:
    0.13 hPa (79 °C)
  • Flash Point:
    147 °C
  • Refractive Index:
    1.428 (40 °C, 589 nm)
  • Density:
    0.89 g/cm3 (20 °C)
  • pH:
    4 (0.2 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together