Order Entry
Poland
ContactUsLinkComponent
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Flammable
Irritant
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.1 %
  • Absorbance (260 nm):
    Max. 0.15
  • Absorbance (270 nm):
    Max. 0.05
  • Absorbance (300 nm):
    Max. 0.02
  • Transmittance (260 nm):
    Min. 70 %
  • Transmittance (270 nm):
    Min. 90 %
  • Transmittance (300 nm):
    Min. 96 %

Specification Test Results

Frequently Bought Together