Order Entry
Poland
ContactUsLinkComponent
Butyl acetate
Butyl acetate
Catalog # PROOCHE131
CAS Number:  
undefined
Butyl acetate
Catalog # PROOCHE131
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :116.16 g/mol
Merck Index:13,01534
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–76 °C
Density:0.875 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
 :GHS02
 :GHS07
CAS Number:123-86-4
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445