Butyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-86-4
:
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445
:116.16 g/mol
Merck Index:14,1535
Boiling Pt:124…126 °C
Melting Pt:–78 °C
Density:0.881
Flash Pt:25 °C (77 °F)
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Related Information
Frequently Bought Together