

Acetone
Catalog # PROOCHE100
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765