Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:200-662-2
:1090
:3,II
:M-Clarity™ quality level = MQ300
:<–20 °C (closed cup)
:01-2119471330-49
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Solubility:(20 °C) soluble
- Melting Point:-95.4 °C
- Molar Mass:58.08 g/mol
- Boiling Point:56.2 °C (1013 hPa)
- Vapor Pressure:233 hPa (20 °C)
- Refractive Index:1.35868 (20 °C)
- Explosion Limit:2.6 - 12.8%(V)
- Density:0.79 g/cm³ (20 °C)
- pH:5 - 6 (395 g/L, H₂O, 20 °C)
- Ignition Point:465 °C DIN 51794
- Water Absorption:1000 g/kg
Frequently Bought Together