Order Entry
Poland
ContactUsLinkComponent
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Irritant
Flammable
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
   Sustainable Options Available
Certificates
undefined
Irritant
Flammable
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.2 % (Coulometric)
  • GC:
    99.9 to 100 % (on anhydrous basis)
  • Color scale:
    ≤10 APHA
  • GC/ECD (as lindane):
    ≤2 ng/l (EOX suitab.)
  • GC/ECD:
    ≤5 ng/l (volatile imp. (as E.Dibromide))
  • Residue after evaporation:
    ≤0.0002 %
  • Acidity (CH3COOH):
    ≤0.002 %

Specification Test Results

Related Information

Frequently Bought Together