Order Entry
Poland
ContactUsLinkComponent
Titanium(IV) oxide, Sigma-Aldrich®
Titanium(IV) oxide, Sigma-Aldrich®
Catalog # SIAL718467-100G
Supplier:  Merck
CAS Number:  
Titanium(IV) oxide, Sigma-Aldrich®
Catalog # SIAL718467-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :79.88 g/mol
Merck Index:13,09549
Boiling Pt:2900 °C (1013 hPa)
Melting Pt:1855 °C
Density:4.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-675-5
CAS Number:13463-67-7
 :1317-70-0
Formula:TiO₂
MDL Number:MFCD00011269