


Dicyklopentadien ≥90% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-73-6
:
Formula:C₁₀H₁₂
MDL Number:MFCD00078246
:132.21 g/mol
Merck Index:14,2739
Boiling Pt:171…173 °C
Melting Pt:12…34 °C
Density:0,982
Flash Pt:26 °C (78 °F)
Storage Temperature:Temperatura otoczenia
UN:2048
ADR:3,III
: w 4-tert-butylopirokatechina
EINECS:201-052-9
:GHS02
:GHS07
:GHS09
Related Information
Frequently Bought Together