
Benzamidine hydrochloride hydrate
Catalog # MOLE12268137-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:174.63 g/mol
EINECS:216-795-4
:GHS07
Storage Temperature:Ambient
CAS Number:206752-36-5
:1670-14-0
Formula:C₆H₅C(=NH)NH₂·HCl·H₂O
MDL Number:MFCD00066285