
Chlorowodorek benzamidyny hydrat, (H₂O 10-14%) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:206752-36-5
:1670-14-0
Formula:C₆H₅C(=NH)NH₂·HCl·H₂O
MDL Number:MFCD00066285
:174.63 g/mol
Melting Pt:82…85 °C
EINECS:216-795-4
:GHS07
Storage Temperature:Temperatura otoczenia
Related Information
Frequently Bought Together