
5-Chloro-2-methoxy-4-methylbenzenesulphonyl chloride ≥97%
Catalog # H26020.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
CAS Number:889939-48-4
:
Formula:C₈H₈Cl₂O₃S
:255.12
Melting Pt:126…128 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
MDL Number:MFCD08445709