Order Entry
Poland
ContactUsLinkComponent
2,5-Difluorobenzenesulphonamide, Sigma-Aldrich®
2,5-Difluorobenzenesulphonamide, Sigma-Aldrich®
Catalog # SIAL553875-1G
Supplier:  Merck
CAS Number:  
2,5-Difluorobenzenesulphonamide, Sigma-Aldrich®
Catalog # SIAL553875-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.17 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:120022-63-1
Formula:C₆H₅F₂NO₂S
MDL Number:MFCD00798115