Order Entry
Poland
ContactUsLinkComponent
3,5-Di-tert-butylaniline ≥97%
Irritant
3,5-Di-tert-butylaniline ≥97%
Catalog # H32768.03
CAS Number:  
Irritant
3,5-Di-tert-butylaniline ≥97%
Catalog # H32768.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.34
Melting Pt:54…57 °C
EINECS:219-173-0
 :GHS07
Storage Temperature:Ambient
CAS Number:2380-36-1
Formula:C₁₄H₂₃N
MDL Number:MFCD00134096