

1,4-Diaminobutane dihydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:333-93-7
:
Formula:C₄H₁₄Cl₂N₂
MDL Number:MFCD00012526
Density:1.31 g/cm³ (20 °C)
ADR:6.1 (8),II
:M-Clarity™ quality level = MQ200
:161.07 g/mol
Storage Temperature:Ambient
EINECS:206-375-9
UN:2928
:GHS06
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:161.07 g/mol
Specification Test Results
Frequently Bought Together