3-(Trifluoromethyl)phenylacetonitrile, Sigma-Aldrich®
Catalog # SIAL199192-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:185.15 g/mol
Boiling Pt:92 °C (1013 hPa)
Density:1.18 g/cm³ (20 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:219-053-8
UN:3276
CAS Number:2338-76-3
Formula:C₉H₆F₃N
MDL Number:MFCD00001913