Tyramine hydrochloride
Catalog # MOLE13456061-25G
Supplier: Molekula
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:173.64
Melting Pt:270…272 °C
EINECS:200-462-5
CAS Number:60-19-5
Formula:C₈H₁₂ClNO
MDL Number:MFCD00012901