Order Entry
Poland
ContactUsLinkComponent
 
Tyramine hydrochloride, Sigma-Aldrich®
  8.18444.0025
 :  Merck
 :  
Tyramine hydrochloride, Sigma-Aldrich®
  8.18444.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    268 - 269 °C
  • Molar Mass:
    173.64 g/mol
  • Synonyms:
    4-(2-Aminoethyl)phenol hydrochloride, Tyramine hydrochloride
  • Hill Formula:
    C8H12ClNO
  • Chemical Formula:
    C8H11NO * HCl

 

 

 :
 :173.64
 :270…272 °C
 :M-Clarity™ quality level = MQ200
 :200-462-5
 :60-19-5
 :C₈H₁₂ClNO
 :MFCD00012901