Order Entry
Poland
ContactUsLinkComponent
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Flammable
Irritant
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Appearance:
    Passes Test
  • Assay (GC):
    ≥99.9 %
  • Color (APHA):
    ≤10
  • Water (H₂O):
    ≤0.02 %
  • Gradient Elution Test (a.u.) - 254 nm:
    ≤0.005
  • Residue after Evaporation:
    ≤3 ppm
  • Titrable Acid (µeq/g):
    ≤8
  • Titrable Base (µeq/g):
    ≤0.6
  • Ultraviolet Absorbance (1.00 cm) - 200 nm:
    ≤0.1
  • Ultraviolet Absorbance (1.00 cm) - 210 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00 cm) - 220 nm:
    ≤0.03
  • Ultraviolet Absorbance (1.00 cm) - 254 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00 cm) - 280 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00 cm) - 350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00 cm) - 400 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00 cm) - UV Cut-off, nm:
    ≤190
  • Filtered through a 0.2 micron filter.:
  • —:

Specification Test Results

Related Information

Frequently Bought Together