Order Entry
Poland
ContactUsLinkComponent
1,3-Dibromoacetone 70% tech.
Irritant
1,3-Dibromoacetone 70% tech.
Catalog # ACRO406550250
CAS Number:  
undefined
Irritant
1,3-Dibromoacetone 70% tech.
Catalog # ACRO406550250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Yellow to dark brown Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥65 %

Specification Test Results

About this item

 :
Boiling Pt:95 °C (20 mmHg)
Melting Pt:28 °C
Density:2,167 g/cm³ (18 °C)
ADR:8,II
 :215.87 g/mol
EINECS:212-430-8
 :GHS07
CAS Number:816-39-7
Formula:BrH₂CC(O)CH₂Br
MDL Number:MFCD00013540