Order Entry
Poland
ContactUsLinkComponent
Dibromomethane stabilised for synthesis, Sigma-Aldrich®
Irritant
Dibromomethane stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Dibromomethane stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    11.7 g/l (15 °C)
  • Melting Point:
    -52.7 °C
  • Molar Mass:
    173.83 g/mol
  • Boiling Point:
    95 - 97 °C (1013 hPa)
  • Vapor Pressure:
    44 hPa (20 °C)
  • Refractive Index:
    1.5419 (20 °C, 589 nm)
  • Density:
    2.49 g/cm3 (20 °C)
  • Synonyms:
    Methylene bromide
  • Hill Formula:
    CH2Br2

Specification Test Results

Related Information

Frequently Bought Together