1,5-Dibromopentane 97%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:111-24-0
- Formula:BrH₂C(CH₂)₃CH₂Br
- MDL Number:MFCD00000268
- Molecular Weight:229.94 g/mol
- Boiling Pt:224 °C (1013 hPa)
- Melting Pt:–34 °C
- Density:1.68 g/cm³
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:203-849-7
2 Options Available Below
Product Details & Documents
- CAS Number:111-24-0
- Formula:BrH₂C(CH₂)₃CH₂Br
- MDL Number:MFCD00000268
- Molecular Weight:229.94 g/mol
- Boiling Pt:224 °C (1013 hPa)
- Melting Pt:–34 °C
- Density:1.68 g/cm³
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:203-849-7
Documents
ACRO15599
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5112 to 1.5132 (20°C, 589 nm)
- GC:≥96.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



