Order Entry
Poland
ContactUsLinkComponent
Silica gel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®
Silica gel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®
Catalog # 1.03804.0001
Supplier:  Merck
CAS Number:  
Silica gel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®
Catalog # 1.03804.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Metal can with PE liner
  • Pk:
    50
  • Seq:
    0050

Specifications

Specification Test Results

  • Water absorption capacity (24 h, 80 % relative humidity):
    ≥ 20 %
  • Loss on drying (140°C, 3 h):
    ≤ 10 %
  • Water Absorption:
    250 g/kg (20 °C)

Specification Test Results

About this item

 :
 :60.08 g/mol
Melting Pt:> 1000 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232