



Sodium chromate tetrahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10034-82-9
:
Formula:CrNa₂O₄·4H₂O
MDL Number:MFCD00149167
:234.03 g/mol
Merck Index:14,8601
Melting Pt:792.2 °C
Density:2.73
EINECS:231-889-5
UN:3288
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS09
Storage Temperature:Ambient
Frequently Bought Together