Order Entry
Poland
ContactUsLinkComponent
di-Sodium tetraborate decahydrate ≥99.5% ACS, Sigma-Aldrich®
Health
di-Sodium tetraborate decahydrate ≥99.5% ACS, Sigma-Aldrich®
Catalog # SIALS9640-25G
Supplier:  Merck
CAS Number:  
Health
di-Sodium tetraborate decahydrate ≥99.5% ACS, Sigma-Aldrich®
Catalog # SIALS9640-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
 :GHS08
CAS Number:1303-96-4
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193