Order Entry
Poland
ContactUsLinkComponent
 
 
 
 
 
(R)-(+)-Limonene for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
 
 
(R)-(+)-Limonene for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    0.02 g/l (25 °C)
  • Melting Point:
    -74 °C
  • Molar Mass:
    136.24 g/mol
  • Boiling Point:
    177 - 178 °C (1013 hPa)
  • Vapor Pressure:
    2.1 hPa (20 °C)
  • Flash Point:
    43 °C DIN 51758
  • Refractive Index:
    1.4722 (20 °C, 589 nm)
  • Explosion Limit:
    0.7 - 6.1 %(V)
  • Density:
    0.84 g/cm3 (20 °C)
  • Ignition Point:
    255 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

 

Frequently Bought Together