Order Entry
Poland
ContactUsLinkComponent
Potassium permanganate 0.002 mol/l (0.01 N) in aqueous solution
Environment
Potassium permanganate 0.002 mol/l (0.01 N) in aqueous solution
Catalog # 85021.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Environment
Potassium permanganate 0.002 mol/l (0.01 N) in aqueous solution
Catalog # 85021.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364