Order Entry
Poland
ContactUsLinkComponent
Sodium carbonate 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium carbonate 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
numer katalogowy 31524.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium carbonate 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
numer katalogowy 31524.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.499 - 0.501 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :105.99 g/mol
EINECS:207-838-8
 :GHS07
Storage Temperature:Ambient
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494