Order Entry
Poland
ContactUsLinkComponent
Hydrogen peroxide 50% stabilised, GPR RECTAPUR®
Hydrogen peroxide 50% stabilised, GPR RECTAPUR®
Catalog # 23620.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrogen peroxide 50% stabilised, GPR RECTAPUR®
Catalog # 23620.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    48 - 51 %
  • Colouration:
    Max. 10 APHA
  • Evaporation residue:
    Max. 0.05 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Total N (Nitrogen):
    Max. 40 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • Ca (Calcium):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Mg (Magnesium):
    Max. 5 ppm

Specification Test Results

About this item

 :
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–40 °C
Density:1.19 g/cm³ (20 °C)
UN:2014
ADR:5.1 (8),II
 :Stabilised with sodium pyrophosphate 30 ppm
 :34.01 g/mol
EINECS:231-765-0
 :GHS03
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333