Hydrogen peroxide 30% (w/w) in water unstabilised ACS
:7722-84-1
:
:H₂O₂
:MFCD00011333
:∼107 °C (1013 hPa)
:–33 °C
:1.11 g/cm³
:Refrigerator
:231-765-0
:2014
:5.1,II
:34.01 g/mol
:GHS03
:GHS05
:GHS07
- Appearance:Clear solution
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Titration with KMnO4:29.0 to 32.0 %
- Chloride (Cl):≤2 ppm
- Nitrate (NO3):≤2 ppm
- Phosphate (PO4):≤2 ppm
- Sulfate (SO4):≤5 ppm
- Ammonium (NH4):≤5 ppm
- Iron (Fe):≤0.5 ppm
- Heavy metals (as Pb):≤1 ppm
- Titrable acid:≤0.0006 meq/g
Frequently Bought Together