Bottles, wide neck, HDPE, round, with screw cap, IP2
Dostawca: Thermo Fisher Scientific
Certyfikaty
Informacje o tym produkcie
Wide-mouth HDPE IP 2 bottles, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant up to +120 °C
- Leakproof when used with Nalgene closures
- Round shape
Safe, durable, economical bottles for the transport and packaging of hazardous goods. Recommended for customers who wish to put together and certify their own packaging system. Extra-wide mouth for simple filling and emptying of liquids and dry substances. The sturdy walls withstand cracking and damage from impact caused by shocks during transport. The bottom of the bottle incorporates IP2 marking, material, volume and registration notch. Evaluated at 15 psi per 49 CFR 173.27 © (2), ICAO Technical Instructions Part 4; 1.1.6 and IATA Dangerous Goods Regulations, Section 5.0.2.9.
Packaging: Bottles and closures are bulk packed in separate polybags within the same carton.
Dane techniczne
- Catalog No:
- NALG312199-0032
- NALG312199-0008
- NALG312199-0016
- NALG312199-0004
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Dane techniczne
Frequently Bought Together