Hydrogen peroxide 30% stabilised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
EINECS:231-765-0
UN:2014
ADR:5.1 (8),II
:34.01 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (manganometric):29.0 - 31.0 %
- Identity:passes test
- Acidity (according to Ph Eur):passes test
- Acidity (according to USP):passes test
- Chloride (Cl):≤ 0.005 %
- Phosphate (PO₄):≤ 0.05 %
- Heavy metals (as Pb):≤ 0.0005 %
- As (Arsenic):≤ 0.0003 %
- Cu (Copper):≤ 0.002 %
- Pb (Lead):≤ 0.001 %
- Pd (Palladium)*:≤ 0.0001 %
- Pt (Platinum)*:≤ 0.0001 %
- Zn (Zinc):≤ 0.002 %
- Residual solvents (ICH Q3C):...excluded by production process
- Organic stabilizers:≤ 0.045 %
- Non volatile matter (105 °C):≤ 0.018 %
- Preservatives:0.015% Na₂H₂P₂O₇; 0.01% H₃PO₄; 0.006% NH₄NO₃
Specification Test Results
Related Information
Frequently Bought Together