Wash bottles, narrow neck, with screw cap
: Brand
PFA, translucent, with ETFE screw cap and FEP spray tube.
- Excellent chemical resistance
- High thermal stability from −200 to 260 °C
- Can be used for less critical applications for which pure PFA is not necessary
With recycled PFA content. Thus, reasonably priced and environmentally friendly.
Frequently Bought Together