
Phosphorus, red 97%, powder, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7723-14-0
:
Formula:P
MDL Number:MFCD00133771
:30.97 g/mol
Melting Pt:590 °C
Density:~ 2,2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-768-7
UN:1338
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Bordeaux to dark brown Powder
- Assay:≥96.0 % (P)
Specification Test Results
Frequently Bought Together