Cesium 99.95+% (trace metals basis)
:7440-46-2
:
:Cs
:MFCD00134037
:132.91
:12,02051
:705 °C (760 mmHg)
:28.5 °C
:1.873 g/cm³
:Ambient
:231-155-4
:1407
:4.3,I
:GHS05
:GHS02
:GHS08
- Appearance:Golden Metal
- Total trace metal impurities:≤500 ppm
- Cesium (Cs):≥99.95 % (metals basis)
Frequently Bought Together