
Sodium tetrachloroaurate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13874-02-7
:
Formula:NaAuCl₄·2H₂O
MDL Number:MFCD00149162
:397.8 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ100
EINECS:239-241-3
UN:3260
:GHS07
Frequently Bought Together