Order Entry
Poland
ContactUsLinkComponent
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Sigma-Aldrich®
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Sigma-Aldrich®
Catalog # SIALN1630-25MG
Supplier:  Merck
CAS Number:  
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Sigma-Aldrich®
Catalog # SIALN1630-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :833.35 g/mol
 :M-Clarity™ quality level = MQ300
EINECS:220-163-3
CAS Number:2646-71-1
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number:MFCD00036263