β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate 97+%
:698999-85-8
:
:C₂₁H₂₇N₇NaO₁₇P₃·xH₂O
:MFCD00150480
:175…178 °C (dec.)
:Freezer
:214-664-6
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Assay:≥97.0 % (NADP, mono-Na) (calc. from enzymatic determination, based on dry weight)
- Water:≤6.0 % (K.F.)
- Ratio of Absorbances:0.80 to 0.86 (A250/A260, pH 7.5)
Frequently Bought Together