(±)-Camphene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-92-5
:
Formula:C₁₀H₁₆
MDL Number:MFCD00066603
:136.24 g/mol
Merck Index:12,01777
Boiling Pt:318 °C (1013 hPa)
Melting Pt:42…45 °C
Density:0.8421 g/cm³ (20 °C)
EINECS:201-234-8
UN:1325
ADR:4.1,III
:M-Clarity™ quality level = MQ100
Flash Pt:34 °C
Frequently Bought Together