Ultrapure water system, Direct-Q® 5, Millipore®
172-5031
: Merck
- Flow rate:Up to 0,6 l/min
- Description:Direct-Q® 5 system
- Environmentally Preferable:
- Operating weight with full tank:29,6 kg
- TOC content:<10 ppb with Quantum EX cartridge (typical value)
- W×D×H:500×260×460 mm (incl. tank)
- Pk:1
For the production of Type I water.
- Automatic recirculation cycles and built-in pressure, heat, conductivity and resistance sensors ensure consistently high water quality
- Warning messages are displayed for maintenance measures
- RS232 interface for easy data retrieval
- With flexible dispensing arm for easy use
- Compact design with small footprint; can be wall or bench mounted
The Direct-Q® 5 water treatment system was developed for scientists who have a daily requirement of 5 to 15 litres of ultrapure water and for whom no pre-treated water of sufficient quality is available. The system combines highest efficiency with low investment and operating costs. It is ideal for preparing buffers for LC analysis, blanks and standard solutions for HPLC as well as a wide range of other analyses and biochemical experiments. In the purifying process, drinking water flows through a pre-treatment pack, a reverse osmosis module, activated carbon, mixed-bed ion exchange resins and a 0,22 µm membrane filter for the final filtration.