Analogue thermo-hygrometers
Supplier: Thies Clima
Certificates
About this item
These combined indicating instruments measure ambient temperature and relative humidity simultaneously.
- Round design
- Black figures on white background
- Two coloured arrows for temperature and humidity
Certifications: Compliant with DIN 50014.
Frequently Bought Together