Order Entry
Poland
ContactUsLinkComponent
Wide horizontal gel electrophoresis system, Owl™ A3-1
Wide horizontal gel electrophoresis system, Owl™ A3-1
Catalog # 730-1112
undefined
Wide horizontal gel electrophoresis system, Owl™ A3-1
Catalog # 730-1112
Specifications
  • Model:
    Owl A3-1
  • Description:
    Wide horizontal gel system
  • Buffer volume:
    4500 ml
  • Gel size W×L:
    230×400 mm
  • Unit W×L×H:
    320×530×110 mm
  • Pk:
    1
Specifications
About this item
The Owl™ A3-1 horizontal gel system can run from 25 to 600 samples on one gel. The gel can be cast in varying lengths to help conserve agarose, when fewer samples need to be run, by utilising a wall comb. The large number of comb slots gives sample and resolving distance flexibility.
  • UV transparent gel tray incorporates gasketed end gates which provide a leak-proof seal without tape
  • Built in levelling ensures the casting of flat, uniformly thick gels
  • Using the specially engineered combs and a multichannel pipette, samples may be loaded directly from a 96-well plate, 8 or 12 at a time
  • Comb slots on the UV transparent gel tray are positioned so that up to ten sample sets of equal lengths can be arranged on the same gel
* Optimal for multi-channel pipetting.
Delivery information: Each system includes buffer chamber, SuperSafe lid with attached power leads, gasketed UVT gel tray and two combs (10- and 14-well, double-sided, 1,0/1,5 mm thick).
Combs
Product Information