Wipes for general applications, Lavette Super
Extra strong hygienic wipes with outstanding performance. Available in five colours.
- 70% viscose, 30% polyester, reusable
- Unique open structure, inhibits growth of micro-organisms and absorbs up to 9 times its own weight
- Excellent resistance, even when wet
- Wipes can be machine washed up to 15 times
- Good resistance to bleach and can be autoclaved at 135 °C for 7 min
Frequently Bought Together