Order Entry
Poland
ContactUsLinkComponent
Base with clamps, flexible
Base with clamps, flexible
Catalog # 241-7510
undefined
Base with clamps, flexible
Catalog # 241-7510

Specifications

  • Length:
    300 mm
  • Pk:
    1

Specifications

About this item

Foot made of die-cast zinc with flexible 300 mm long three-finger clamp, nickel plated brass.

  • Safety base, 85 mm diameter, for firm immovable positioning
  • Clamp spanning width 0 to 40 mm
  • The fingers of the clamp are covered with a plastic coating to protect the items clamped