Order Entry
Poland
ContactUsLinkComponent
 
Sedimentation cones, Imhoff type, Nalgene®
  532-0101
   Low Manufacturing Impact
undefined
Sedimentation cones, Imhoff type, Nalgene®
  532-0101
   Low Manufacturing Impact

 

  • Description:
    Imhoff cone 1000 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    1

 

 

PC with PP autoclavable stopper.

  • As transparent as glass
  • Easy draining via screw plug
  • Leakproof closure, closure design allows user to read graduations to zero

Graduation: 0 to 1 ml/0,1; 1 to 10 ml/0,5; 10 to 20 ml/1; 20 to 40 ml/2; 40 to 100 ml/10; 100 to 500/50; 500 to 1000 ml/100.