Order Entry
Poland
Orders LinkContactUsLinkComponent
Bottle top tilt measures
Supplier:   KARTELL
Bottle top tilt measures
Supplier:  KARTELL
Certificates
Bottle top tilt measures
Bottle top tilt measures
6 Options Available Below
Product Details & Documents
PE (bottle and tube), PMP (measuring cup).
  • Repetitive dispensing of the same volume of liquid
  • When the bottle pressure is released only the desired volume remains
The 25 /50 ml measuring cup slides up and down the tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
Colour: Transparent /Red
Product Family Options
Product Information
  • Capacity
    Colour
    Availability
    Price
  • 25 /250 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 42,20 zł
    42,20 zł
    1 * 1 SZT
       42,20 zł
  • 25 /500 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 45,60 zł
    45,60 zł
    1 * 1 SZT
       45,60 zł
  • 25 /1000 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 51,30 zł
    51,30 zł
    1 * 1 SZT
       51,30 zł
  • 50 /250 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 52,90 zł
    52,90 zł
    1 * 1 SZT
       52,90 zł
  • 50 /500 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 56,60 zł
    56,60 zł
    1 * 1 SZT
       56,60 zł
  • 50 /1000 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 70,10 zł
    70,10 zł
    1 * 1 SZT
       70,10 zł