Order Entry
Poland
Orders LinkContactUsLinkComponent
Top tilt measures
Dostawca:   Lenz Laborglas
Top tilt measures
Dostawca:  Lenz Laborglas
Certyfikaty
Top tilt measures
Top tilt measures
9 dostępnych opcji poniżej
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These tilt measures with ground joint cone NS 29/32 can fit flasks - see 215-1117 and 215-1118.
Product Family Options
Informacje o produkcie